CAS RN 2585-24-2
IUPAC Name:
4-nitro-N-propylbenzamide
Synonyms:
4-nitro-N-propylbenzamide
Molecular Weight (g/mol):
208.22
Molecular Formula:
C10H12N2O3
InChi Key:
MDRVSZJDSUDYGP-UHFFFAOYSA-N
SMILES:
CCCNC(=O)C1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-Nitro-N-n-propylbenzamide, 97%, Thermo Scientific™
CAS: 2585-24-2 Molecular Formula: C10H12N2O3 Molecular Weight (g/mol): 208.217 MDL Number: MFCD00024716 InChI Key: MDRVSZJDSUDYGP-UHFFFAOYSA-N PubChem CID: 314664 IUPAC Name: 4-nitro-N-propylbenzamide SMILES: CCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]