accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2586-62-1

IUPAC Name: 1-bromo-2-methylnaphthalene
Synonyms: 1-bromo-2-methylnaphthalene
Molecular Weight (g/mol): 221.1
Molecular Formula: C11H9Br
InChi Key: CMIMBQIBIZZZHQ-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=C1Br
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (4)