accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2588-04-7

IUPAC Name: O,O-diethyl {[(ethanesulfonyl)methyl]sulfanyl}phosphonothioate
Synonyms: phorate sulfone
Molecular Weight (g/mol): 292.36
Molecular Formula: C7H17O4PS3
InChi Key: YVPSNUIHHFTTRL-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCS(=O)(=O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results