accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25912-91-8

IUPAC Name: 2-dodecyl-4-methylphenol
Synonyms: 2-dodecyl-4-methylphenol
Molecular Weight (g/mol): 276.46
Molecular Formula: C19H32O
InChi Key: OCDBOZHPSDAPFB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=C(O)C=CC(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results