CAS RN 259209-20-6
IUPAC Name:
(5-fluoro-2-hydroxyphenyl)boronic acid
Synonyms:
5-fluoro-2-hydroxyphenylboronic acid
Molecular Weight (g/mol):
155.92
Molecular Formula:
C6H6BFO3
InChi Key:
TWDQSJDFXUMAOI-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(O)C=CC(F)=C1