accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CSiOHNOHO

CAS RN 259212-61-8

IUPAC Name: 4-[(tert-butyldiphenylsilyl)oxy]pyrrolidine-2-carboxylic acid
Synonyms: 4-[(tert-butyldiphenylsilyl)oxy]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 369.54
Molecular Formula: C21H27NO3Si
InChi Key: LHSRURYOCDIZEW-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](OC1CNC(C1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results