accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CCH3CH3NCH3OOH3CNH3COOH3CCH3Co(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 259259-80-8

IUPAC Name: λ²-cobalt(2+) 3-[({2-[(2-acetyl-3-oxidobut-2-en-1-ylidene)amino]-1,2-bis(2,4,6-trimethylphenyl)ethyl}imino)methyl]-4-oxopent-2-en-2-olate
Synonyms: λ²-cobalt(2+) 3-[({2-[(2-acetyl-3-oxidobut-2-en-1-ylidene)amino]-1,2-bis(2,4,6-trimethylphenyl)ethyl}imino)methyl]-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 573.6
Molecular Formula: C32H38CoN2O4
InChi Key: PHCQQLMRNZRDJA-UHFFFAOYNA-L
SMILES: [Co++].CC([O-])=C(C=NC(C(N=CC(=C(C)[O-])C(C)=O)C1=C(C)C=C(C)C=C1C)C1=C(C)C=C(C)C=C1C)C(C)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results