accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSNNClClCl

CAS RN 2593-15-9

IUPAC Name: 5-ethoxy-3-(trichloromethyl)-1,2,4-thiadiazole
Synonyms: koban etridazole
Molecular Weight (g/mol): 247.52
Molecular Formula: C5H5Cl3N2OS
InChi Key: KQTVWCSONPJJPE-UHFFFAOYSA-N
SMILES: CCOC1=NC(=NS1)C(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results