accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25932-11-0

IUPAC Name: sodium benzenesulfinate dihydrate
Synonyms: sodium benzenesulfinate dihydrate
Molecular Weight (g/mol): 200.18
Molecular Formula: C6H9NaO4S
InChi Key: MYXJYAIKMQJHIB-UHFFFAOYSA-M
SMILES: O.O.[Na+].[O-]S(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)