accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25979-07-1

IUPAC Name: tert-butyldichlorophosphane
Synonyms: tert-butyldichlorophosphane
Molecular Weight (g/mol): 158.99
Molecular Formula: C4H9Cl2P
InChi Key: NMJASRUOIRRDSX-UHFFFAOYSA-N
SMILES: CC(C)(C)P(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)