accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2N(S)NH2ONH(S)NH2OHN(S)NH2OHO

CAS RN 25988-63-0

IUPAC Name: (2S)-6-amino-2-[(2S)-6-amino-2-[(2S)-2,6-diaminohexanamido]hexanamido]hexanoic acid
Synonyms: polylysine hbr
Molecular Weight (g/mol): 402.54
Molecular Formula: C18H38N6O4
InChi Key: WBSCNDJQPKSPII-KKUMJFAQSA-N
SMILES: NCCCC[C@H](N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results