accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2603-10-3

IUPAC Name: methyl N-phenylcarbamate
Synonyms: methyl N-phenylcarbamate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: IAGUPODHENSJEZ-UHFFFAOYSA-N
SMILES: COC(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results