accessibility menu, dialog, popup

UserName

Viewing profile as user:
IFFFCl

CAS RN 260355-20-2

IUPAC Name: 1-chloro-4-iodo-2-(trifluoromethyl)benzene
Synonyms: 1-chloro-4-iodo-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 306.45
Molecular Formula: C7H3ClF3I
InChi Key: MCIFWVOVVPVBRJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=CC(I)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results