accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26046-90-2

H3CSe(R)NH2OHO

CAS RN 26046-90-2

IUPAC Name: (2R)-2-amino-3-(methylselanyl)propanoic acid
Synonyms: methylselenocysteine DL-se-methylselenocysteine SeMSC se-methyl-seleno-L-cysteine l-se-methylselenocysteine SeMCys se-methyl-selenocysteine selenomethylselenocysteine selenium methyl cysteine C4H8NO2se
Molecular Weight (g/mol): 182.09
Molecular Formula: C4H9NO2Se
InChi Key: XDSSPSLGNGIIHP-VKHMYHEASA-N
SMILES: C[Se]C[C@H](N)C(O)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)