accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26060-56-0

IUPAC Name: (E)-(1-phenylethylidene)amino benzoate
Synonyms: (E)-(1-phenylethylidene)amino benzoate
Molecular Weight (g/mol): 239.27
Molecular Formula: C15H13NO2
InChi Key: KLJLQTJYNGGTIU-FOWTUZBSSA-N
SMILES: C\C(=N/OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results