accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26107-82-4

IUPAC Name: 3,5-bis(trifluoromethyl)benzohydrazide
Synonyms: 3,5-bis(trifluoromethyl)benzohydrazide
Molecular Weight (g/mol): 272.15
Molecular Formula: C9H6F6N2O
InChi Key: GBBRFBSFWKFTMZ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results