accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaOSOOONNHSOOOSOOO(Z)

CAS RN 2611-82-7

IUPAC Name: trisodium (8Z)-7-oxo-8-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-7,8-dihydronaphthalene-1,3-disulfonate
Synonyms: acid red 18
Molecular Weight (g/mol): 604.46
Molecular Formula: C20H11N2Na3O10S3
InChi Key: IVNZBWNBYXERPK-DZGBHZPSSA-K
SMILES: [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (2)
Quantity 
  • (2)
  • (4)
  • (1)
Color 
  • (2)
  • (3)
  • (1)
Grade 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)