accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2613-26-5

IUPAC Name: 1-nitro-3-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-nitro-3-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 249.16
Molecular Formula: C6H4F5NO2S
InChi Key: FSTNQYCPXJMFMT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results