accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOSFFFFF

CAS RN 2613-27-6

IUPAC Name: 1-nitro-4-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-nitro-4-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 249.16
Molecular Formula: C6H4F5NO2S
InChi Key: AGNCKMHGYZKMLN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)S(F)(F)(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results