accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26134-62-3

IUPAC Name: Lithium nitride
Synonyms: trilithium nitride|Li3N
Molecular Weight (g/mol): 34.83
Molecular Formula: Li3N
InChi Key: IDBFBDSKYCUNPW-UHFFFAOYSA-N
SMILES: [Li]N([Li])[Li]
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)