accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 261360-77-4

IUPAC Name: (2R)-3-(3-bromophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: (2R)-3-(3-bromophenyl)-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 343.2
Molecular Formula: C14H17BrNO4
InChi Key: FBUDYESOPLBQIR-LLVKDONJSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=CC(Br)=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)