accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNOHO

CAS RN 261380-28-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(2-cyanophenyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(2-cyanophenyl)propanoic acid
Molecular Weight (g/mol): 290.32
Molecular Formula: C15H18N2O4
InChi Key: AJHPGXZOIAYYDW-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C#N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)