accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26143-11-3

IUPAC Name: pentaethoxytungsten
Synonyms: pentaethoxytungsten
Molecular Weight (g/mol): 409.15
Molecular Formula: C10H25O5W
InChi Key: SRGJJDPFERQMIL-UHFFFAOYSA-N
SMILES: CCO[W](OCC)(OCC)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)