accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHO

CAS RN 26153-38-8

IUPAC Name: 3,5-dihydroxybenzaldehyde
Synonyms: 3,5-dihydroxybenzaldehyde
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: HAQLHRYUDBKTJG-UHFFFAOYSA-N
SMILES: OC1=CC(C=O)=CC(O)=C1
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)