accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 261633-71-0

IUPAC Name: 1-(2,3-dihydro-1,4-benzodioxin-5-yl)methanamine
Synonyms: 1-(2,3-dihydro-1,4-benzodioxin-5-yl)methanamine
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: OKRXRKUGZVMPKX-UHFFFAOYSA-N
SMILES: NCC1=C2OCCOC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results