accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 261635-93-2

IUPAC Name: 2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid
Synonyms: 2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: ODWJFUQRQUCRJZ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(=N1)C(F)(F)F)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results