accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 261723-32-4

IUPAC Name: (3-bromo-2-fluorophenyl)methanol
Synonyms: (3-bromo-2-fluorophenyl)methanol
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: LIZLYZVAYZQVPG-UHFFFAOYSA-N
SMILES: OCC1=C(F)C(Br)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results