accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 261763-34-2

IUPAC Name: 2,6-difluoro-3-methylbenzaldehyde
Synonyms: 2,6-difluoro-3-methylbenzaldehyde
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: MXZXYEFIOADJMW-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(C=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results