accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 261763-46-6

IUPAC Name: 2,6-difluoro-3-methylphenol
Synonyms: 2,6-difluoro-3-methylphenol
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: YKCLAAQQWYPEDW-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results