accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26177-86-6

IUPAC Name: [(2,3,4,6-tetrahydroxy-5-oxohexyl)oxy]phosphonic acid disodium
Synonyms: fructose-6-phosphate disodium
Molecular Weight (g/mol): 306.11
Molecular Formula: C6H13Na2O9P
InChi Key: VAPLWAFNCWDFQF-UHFFFAOYNA-N
SMILES: [Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results