accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFOOH

CAS RN 261945-09-9

IUPAC Name: 3,5-difluoro-4-(trifluoromethyl)benzoic acid
Synonyms: 3,5-difluoro-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 226.1
Molecular Formula: C8H3F5O2
InChi Key: MKESWMDKZFRERU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(C(F)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results