CAS RN 261951-68-2
CAS RN 261951-68-2
IUPAC Name:
1-(4-fluoro-3-methylphenyl)methanamine
Synonyms:
1-(4-fluoro-3-methylphenyl)methanamine
Molecular Weight (g/mol):
139.17
Molecular Formula:
C8H10FN
InChi Key:
PQXWYQZQCWWMDQ-UHFFFAOYSA-N
SMILES:
CC1=CC(CN)=CC=C1F
1
–
1
of
1
results
1
Sigma Aldrich 4-Fluoro-3-methylbenzylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.