accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrFFF

CAS RN 261952-20-9

IUPAC Name: 1-bromo-4-methyl-2-(trifluoromethyl)benzene
Synonyms: 1-bromo-4-methyl-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.04
Molecular Formula: C8H6BrF3
InChi Key: WXXDBPIOFOYRLP-UHFFFAOYSA-N
SMILES: CC1=CC(=C(Br)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results