accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 262-89-5

IUPAC Name: (2Z,10Z)-tricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),2,4,6,8,10,12,14-octaene
Synonyms: (2Z,10Z)-tricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),2,4,6,8,10,12,14-octaene
Molecular Weight (g/mol): 204.27
Molecular Formula: C16H12
InChi Key: VGQWCQNHAMVTJY-RKUAQPHYSA-N
SMILES: C1=CC=C2\C=C/C3=CC=CC=C3\C=C/C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results