accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2620-44-2

IUPAC Name: 5-nitro-2H-1,3-benzodioxole
Synonyms: 5-nitro-2H-1,3-benzodioxole
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: SNWQAKNKGGOVMO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OCOC2=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)