accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 2620-76-0

IUPAC Name: 2-(4-bromophenyl)-1-phenyl-1H-1,3-benzodiazole
Synonyms: 2-(4-bromophenyl)-1-phenyl-1,3-benzodiazole
Molecular Weight (g/mol): 349.23
Molecular Formula: C19H13BrN2
InChi Key: DXRLALXPCIOIDK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)