accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26215-14-5

IUPAC Name: 7-amino-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 7-amino-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: RUZXDTHZHJTTRO-UHFFFAOYSA-N
SMILES: NC1=CC=C2NC(=O)COC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results