accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26215-90-7

IUPAC Name: tridecan-4-one
Synonyms: tridecan-4-one
Molecular Weight (g/mol): 198.35
Molecular Formula: C13H26O
InChi Key: JWSRUPAFLPWLNO-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)CCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results