accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2622-74-4

IUPAC Name: 2-(4-bromophenyl)-1H-1,3-benzodiazole
Synonyms: 2-(4-bromophenyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 273.13
Molecular Formula: C13H9BrN2
InChi Key: YRWMGOSKROWAIT-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)