accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2624-44-4

IUPAC Name: 2,5-dihydroxybenzene-1-sulfonic acid; diethylamine
Synonyms: etamsylate 酚磺乙胺
Molecular Weight (g/mol): 263.31
Molecular Formula: C10H17NO5S
InChi Key: HBGOLJKPSFNJSD-UHFFFAOYSA-N
SMILES: CCNCC.OC1=CC=C(O)C(=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results