accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClOOHCl

CAS RN 262423-77-8

IUPAC Name: 2,4-dichloropyridine-3-carboxylic acid
Synonyms: 2,4-dichloropyridine-3-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: ZFGZNVSNOJPGDV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CN=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)