accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPOHOHO

CAS RN 2627-35-2

IUPAC Name: (dodecyloxy)phosphonic acid
Synonyms: dodecyloxyphosphonic acid
Molecular Weight (g/mol): 266.32
Molecular Formula: C12H27O4P
InChi Key: TVACALAUIQMRDF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOP(O)(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results