accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3COSiCH3CH3CH2H2C

CAS RN 2627-95-4

IUPAC Name: ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane
Synonyms: ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane
Molecular Weight (g/mol): 186.4
Molecular Formula: C8H18OSi2
InChi Key: BITPLIXHRASDQB-UHFFFAOYSA-N
SMILES: C[Si](C)(O[Si](C)(C)C=C)C=C
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (2)
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (8)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (6)