accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2628-16-2

IUPAC Name: 4-ethenylphenyl acetate
Synonyms: phenol, 4-ethenyl-, acetate
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: JAMNSIXSLVPNLC-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)