accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHN

CAS RN 262861-81-4

IUPAC Name: 9-(4-ethynylphenyl)-9H-carbazole
Synonyms: 9-(4-ethynylphenyl)carbazole
Molecular Weight (g/mol): 267.33
Molecular Formula: C20H13N
InChi Key: JJIQKIUIIVFHAN-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results