accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 262862-97-5

IUPAC Name: [4-(phenoxymethyl)phenyl]methanol
Synonyms: [4-(phenoxymethyl)phenyl]methanol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: MTQHORUPINPVTP-UHFFFAOYSA-N
SMILES: OCC1=CC=C(COC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results