accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 263021-30-3

IUPAC Name: 4-{[(tert-butoxy)carbonyl](methyl)amino}benzoic acid
Synonyms: 4-[(tert-butoxycarbonyl)(methyl)amino]benzoic acid
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: VRQKKKZQZSNRKD-UHFFFAOYSA-N
SMILES: CN(C(=O)OC(C)(C)C)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results