accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2633-66-1

IUPAC Name: bromo(3,4,5-trimethylphenyl)magnesium
Synonyms: bromo(3,4,5-trimethylphenyl)magnesium
Molecular Weight (g/mol): 223.4
Molecular Formula: C9H11BrMg
InChi Key: VZMGGVXGYWKQPB-UHFFFAOYSA-M
SMILES: CC1=CC([Mg]Br)=CC(C)=C1C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results