accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 263349-22-0

IUPAC Name: 3-(pyrimidin-2-yl)benzaldehyde
Synonyms: 3-(pyrimidin-2-yl)benzaldehyde
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: ZBAZYPXXIVHUFO-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results