accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26346-85-0

IUPAC Name: 2-bromo-1-(2,4-dimethylphenyl)ethan-1-one
Synonyms: 2-bromo-1-(2,4-dimethylphenyl)ethanone
Molecular Weight (g/mol): 227.1
Molecular Formula: C10H11BrO
InChi Key: GSCCVWPVPFIRJP-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results